Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIA1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | DIA1 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
DIA1 Polyclonal specifically detects DIA1 in Human samples. It is validated for Western Blot.Specifications
DIA1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
C3orf58, chromosome 3 open reading frame 58, DIA1 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of human DIA1 (NP_001127942.1). Peptide sequence RHATWRGTSGGLLHDPPSEIAKDGRLEALLDECANPKKRYGRFQAAKELR | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
205428 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title