Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIABLO Mouse anti-Human, Clone: 79-1-83, Abnova™

Mouse monoclonal antibody raised against human DIABLO.

Manufacturer:  Abnova Corporation MAB12589

Catalog No. 89-934-043


Add to cart

Description

Description

This gene encodes an inhibitor of apoptosis protein (IAP)-binding protein. The encoded mitochondrial protein enters the cytosol when cells undergo apoptosis, and it moderates the caspase inhibition of IAPs. Multiple polyadenylation sites have been found for this gene. Four alternatively spliced transcript variants have been described for this gene, with two of them encoding different isoforms and the other two probably not encoding a protein. [provided by RefSeq]

Specifications

Specifications

DIABLO
79-1-83
Mouse monoclonal antibody raised against human DIABLO.
In PBS, pH 7.2 (50% glycerol, 0.09% sodium azide).
DIABLO-S/FLJ10537/FLJ25049/SMAC/SMAC3
Mouse
IgG2b
50 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
DIABLO
Recombinant protein corresponding to full length human DIABLO.
Protein G purification
RUO
Primary
56616
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit