Learn More
DIABLO Rabbit anti-Human, Rat, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against recombinant human DIABLO.
Supplier: Abnova Corporation PAB29316
Description
This gene encodes an inhibitor of apoptosis protein (IAP)-binding protein. The encoded mitochondrial protein enters the cytosol when cells undergo apoptosis, and it moderates the caspase inhibition of IAPs. Multiple polyadenylation sites have been found for this gene. Four alternatively spliced transcript variants have been described for this gene, with two of them encoding different isoforms and the other two probably not encoding a protein. [provided by RefSeq]
Sequence: AVYTLTSLYRQYTSLLGKMNSEEEDEVWQVIIGARAEMTSKHQEYLKLETTWMTAVGLSEMAAEAAYQTGADQASITARNHIQLVKLQVEEVHQLSRKAETKLAEAQIEELRQKTQEEGSpecifications
DIABLO | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant human DIABLO. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
DIABLO | |
Recombinant protein corresponding to amino acids of human DIABLO. | |
100 μL | |
Primary | |
Human, Rat | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:20-1:50) Western Blot (1:100-1:250) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
DIABLO-S/FLJ10537/FLJ25049/SMAC/SMAC3 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
56616 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.