Alfa Aesar™ Diacetato[1,3-bis(diphenylphosphino)propane]palladium(II), Pd 16.7%

Manufacturer: Alfa Aesar™ 4637003

Quantity 1g

 View more versions of this product

Catalog No. AA4637003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 149796-59-8
    Formula Weight 636.96
    MDL Number MFCD14155705
    Synonym Pd(OAc)2(dppp)
    Chemical Name or Material Diacetato[1,3-bis(diphenylphosphino)propane]palladium(II)

    Specifications

    Formula Weight 636.96
    Physical Form Crystalline Solid
    MDL Number MFCD14155705
    CAS 149796-59-8
    Color Cream
    Sensitivity Moisture Sensitive
    Concentration Pd 16.7%
    Synonym Pd(OAc)2(dppp)
    Quantity 1g
    Chemical Name or Material Diacetato[1,3-bis(diphenylphosphino)propane]palladium(II)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit