Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diacetin, Spectrum™

Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Amber Glass Bottle
25395-31-7
C7H12O5
176.17
UXDDRFCJKNROTO-UHFFFAOYNA-N
1-(acetyloxy)-3-hydroxypropan-2-yl acetate
CC(=O)OCC(CO)OC(C)=O
25395-31-7
C7H12O5
176.17
UXDDRFCJKNROTO-UHFFFAOYNA-N
1-(acetyloxy)-3-hydroxypropan-2-yl acetate
CC(=O)OCC(CO)OC(C)=O

D3159, 25395-31-7, C7H12O5

Spectrum™ Diacetin
Packaging Amber Glass Bottle
Quantity 100 g
CAS Max % 1