Learn More
Diaion™ CR11(Na), chelating resin, highly porous type, 0.5 mmole/ml on poly(styrene-divinylbenzene), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 046544A1
Description
Hardness removal in brine, selective transitional metal removal
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
355-1180μm Beads | |
Highly porous type, 0.5mmole/mL on poly(styrene-divinylbenzene) | |
Odorless | |
1 kg |
Diaion™ CR11 (Na), chelating resin | |
74350-44-0 | |
355 to 1180μm |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only