Alfa Aesar™ Diaion™ CRB03(OH), chelating resin, highly porous type, 0.7 meq/ml on poly(styrene-divinylbenzene)
Boron, arsenic, gallium, and selenium removal
Manufacturer: Alfa Aesar™ 46861A1
Quantity 1kg
 View more versions of this product
Catalog No. AA46861A1
  Check Availability
Add to Cart
Related Products
Product Content Feedback