Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diaion™ HP21, synthetic adsorbent resin, highly porous type, 40g/L on PS-DVB, P.R. 110 angstroms

Amino acid separations, small molecules in medium hydrophobicity in polar solvents

Manufacturer:  Alfa Aesar™ 46897A1

Quantity 1kg

 View more versions of this product

Catalog No. AA46897A1


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

9052-95-3
MFCD00148061
Diaion™ HP21, synthetic adsorbent resin, Highly porous type, 40g/L on PS-DVB

Specifications

9052-95-3
250μm, P.R. 110Å
1.05g/mL
White to pale yellow
640m2/g
Odorless
P.V. 1.3Ml/G Beads
MFCD00148061
1kg
Diaion™ HP21, synthetic adsorbent resin, Highly porous type, 40g/L on PS-DVB
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit