Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diaion™ HP2MGL, synthetic adsorbent resin, highly porous type, 10g/L on polymethacrylate, P.R. 240 angstroms

Catalog No. AA46860A1
Click to view available options
Quantity:
1 kg

CAS: 25777-71-3 | C15H22O6 | 298.335 g/mol

Amino acids separations, small molecules with high hydrophobicity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25777-71-3
Molecular Formula C15H22O6
Molecular Weight (g/mol) 298.335
InChI Key XBDAKYZJVVYINU-UHFFFAOYSA-N
Synonym bio acrylic resin, methyl 2-methylprop-2-enoate; 2-2-methylprop-2-enoyloxy ethyl 2-methylprop-2-enoate, unii-eg97988m5q, 2-propenoic acid, 2-methyl-, 1,2-ethanediyl ester, polymer with methyl 2-methyl-2-propenoate, methyl methacrylate, ethylene glycol dimethacrylate polymer, methyl methacrylate, polymer with ethylene glycol dimethacrylate, 2-propenoic acid, 2-methyl-, 1,1'-1,2-ethanediyl ester, polymer with methyl 2-methyl-2-propenoate, glycol dimethacrylate; methyl methacrylate, poly methyl methacrylate-co-ethylene glycol dimethacrylate
PubChem CID 3080735
IUPAC Name methyl 2-methylprop-2-enoate;2-(2-methylprop-2-enoyloxy)ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC.CC(=C)C(=O)OCCOC(=O)C(=C)C

Specifications

Physical Form 300μm Beads,S.A. 570m
Odor Odorless
Particle Size 300μm, P.R. 240Å
Density 1.09 g/mL
Color White to Yellow
Quantity 1 kg
Surface Area 570m2/g
Chemical Name or Material Diaion™ HP2MGL, synthetic adsorbent resin, Highly porous type, 10g/L on polymethacrylate
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.