Learn More
Diaion™ HPA25(Cl), ion exchange resin, highly cross-linked, strongly basic highly porous type I, 0.5 meq/ml on PS-DVB, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 046691A1
Description
Enzyme immobilization, catalysis, used in high oxidative environments
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
250μm Beads | |
Highly cross-linked, strongly basic highly porous type I, 0.5meq/mL on PS-DVB | |
250μm |
Diaion™ HPA25 (Cl), ion exchange resin | |
82643-04-7 | |
1 kg |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only