Learn More
Diaion™ PA308(Cl), ion exchange resin, 4% cross-linked, strongly basic porous type I, 1.0 meq/ml on PS-DVB, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 046990A1
Description
Sugar decolorization
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
300-1180μm Beads | |
4% cross-linked, strongly basic porous type I, 1.0meq/mL on PS-DVB | |
White to Yellow |
Diaion™ PA308 (Cl), ion exchange resin | |
300 to 1180μm | |
1 kg |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures