Learn More
Alfa Aesar™ Diaion™ SA11A(Cl), ion exchange resin, lightly cross-linked, strongly basic gel type I, 0.85 meq/ml on PS-DVB
Decolorization in sugar solutions, metal recovery, pharmaceuticals
Manufacturer:  Alfa Aesar™ 46980A1
Quantity 1kg
 View more versions of this product
Catalog No. AA46980A1
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
Diaion™ SA11A (Cl), ion exchange resin, Lightly cross-linked, strongly basic gel type I, 0.85meq/mL on PS-DVB
Specifications
Odorless
Beads
Diaion™ SA11A (Cl), ion exchange resin, Lightly cross-linked, strongly basic gel type I, 0.85meq/mL on PS-DVB
300 to 1180μm
1kg