Alfa Aesar™ Diaion™ UBK550(Na), ion exchange fractionation resin, 8% x-linked, strongly acidic gel type, 1.9meq/mL on PS-DVB
Chromatographic oligosaccharide sugar separations, sugar alcohol separations
Manufacturer: Alfa Aesar™ 46609A1
Quantity 1kg
 View more versions of this product
Catalog No. AA46609A1
  Check Availability
Add to Cart
    Chemical Identifiers
    Chemical Name or Material Diaion™ UBK550 (Na), ion exchange fractionation resin, 8% cross-linked, strongly acidic gel type, 1.9meq/mL on PS-DVB
    Specifications
    Quantity 1kg
    Particle Size 220μm
    Physical Form Beads
    Chemical Name or Material Diaion™ UBK550 (Na), ion exchange fractionation resin, 8% cross-linked, strongly acidic gel type, 1.9meq/mL on PS-DVB
  • Description & Specifications
Related Products
Product Content Feedback