Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diaion™ WA30, ion exchange resin, weakly basic porous type, 1.5 meq/ml on poly(styrene-divinylbenzene), Thermo Scientific Chemicals

Catalog Number aa46592a1
Click to view available options
Quantity:
1 kg
65405-58-5
C21H25N
291.438
OTCJDTFYONDHFM-UHFFFAOYSA-N
divinylbenzene, vinylbenzyl dimethylamine polymer, benzenemethanamine, ar-ethenyl-n,n-dimethyl-, polymer with diethenylbenzene, 2-ethenylphenyl methyl dimethylamine; divinylbenzene, 1,2-bis ethenyl benzene; 1-2-ethenylphenyl-n,n-dimethylmethanamine
6455131
1,2-bis(ethenyl)benzene;1-(2-ethenylphenyl)-N,N-dimethylmethanamine
CN(C)CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1C=C
65405-58-5
C21H25N
291.438
OTCJDTFYONDHFM-UHFFFAOYSA-N
divinylbenzene, vinylbenzyl dimethylamine polymer, benzenemethanamine, ar-ethenyl-n,n-dimethyl-, polymer with diethenylbenzene, 2-ethenylphenyl methyl dimethylamine; divinylbenzene, 1,2-bis ethenyl benzene; 1-2-ethenylphenyl-n,n-dimethylmethanamine
6455131
1,2-bis(ethenyl)benzene;1-(2-ethenylphenyl)-N,N-dimethylmethanamine
CN(C)CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1C=C

 

Water treatment, refining of starch syrup and glucose, refining of beet sugar

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form 300-1180μm Beads
Odor Odorless
Particle Size 300 to 1180μm
Color Yellow
Quantity 1 kg
Chemical Name or Material Diaion™ WA30, ion exchange resin, Weakly basic porous type, 1.5meq/mL on poly(styrene-divinylbenzene)
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit