Learn More
Diaion™ WK100(H), ion exchange resin, weakly acidic porous type, 2.9 meq/ml on polymethacrylate, high reaction rate, Thermo Scientific Chemicals
Supplier:  Thermo Scientific Chemicals 046647A1
Catalog No. AA46647A1
  • $125.65 / Each of 1
    Save 16%
    Reg : $149.00
    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Order in Bulk
Request bulk or custom formats
Add to Cart
Description
Description
Pharmaceuticals, dealkalination, metal recovery, antibiotics, sucrose purifications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
300-1180μm Beads
Weakly acidic porous type, 2.9meq/mL on polymethacrylate, high reaction rate
White
Diaion™ WK100 (H), ion exchange resin
300 to 1180μm
1 kg
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures