Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diallyldibutyltin

Click to view available options
Quantity:
0.5g
2g
10g
15336-98-8
C14H28Sn
315.088
MFCD00014944
LCGVYMRFNOWPGQ-UHFFFAOYSA-N
diallyldibutyltin,diallyldibutylstannane,dibutylbis prop-2-en-1-yl stannane,diallyl dibutyl stannane,diallyldi-n-butyltin,acmc-1bvvj,dibutyl-bis prop-2-enyl stannane
84873
dibutyl-bis(prop-2-enyl)stannane
CCCC[Sn](CCCC)(CC=C)CC=C
15336-98-8
C14H28Sn
315.088
MFCD00014944
LCGVYMRFNOWPGQ-UHFFFAOYSA-N
diallyldibutyltin,diallyldibutylstannane,dibutylbis prop-2-en-1-yl stannane,diallyl dibutyl stannane,diallyldi-n-butyltin,acmc-1bvvj,dibutyl-bis prop-2-enyl stannane
84873
dibutyl-bis(prop-2-enyl)stannane
CCCC[Sn](CCCC)(CC=C)CC=C
Quantity 0.5g
Boiling Point 81°C (0.2mmHg)
Sensitivity Air and moisture sensitive
Density 1.1002 g/mL
Refractive Index 1.5
UN Number UN2788
Solubility Information Insoluble in water.
Formula Weight 315.07
Physical Form Liquid
Chemical Name or Material Diallyldibutyltin

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

EINECSNumber : 239-368-4

TSCA : No

Recommended Storage : Ambient temperatures