Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diallylmethylamine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.7890g/mL |
Boiling Point | 111.0°C |
Flash Point | 7°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Quantity | 5 g |
Specific Gravity | 0.789 |
Synonym | 1,4-bis glycidyloxy benzene, hydroquinone diglycidyl ether, 2-4-oxiran-2-ylmethoxy phenoxymethyl oxirane, hydroquinone 1,4-diglycidyl ether, oxirane, 2,2'-1,4-phenylenebis oxymethylene bis, 2,2'-1,4-phenylenebis oxymethylene dioxirane, 2-4-oxiran-2-ylmethoxy phenoxy methyl oxirane, 1,4-bis oxiran-2-ylmethoxy benzene, acmc-20ap2z |
InChI Key | FSYPIGPPWAJCJG-UHFFFAOYNA-N |
SMILES | C1C(O1)COC2=CC=C(C=C2)OCC3CO3 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Highly flammable liquid and vapor. Causes severe skin burns and eye damage. |
Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call |
EINECSNumber | 219-354-4 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction