Learn More
Spectrum™ Dialysis Tubing Closures: Spectra/Por™ Standard Type, for RC Tubing only
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Manufacturer:  Spectrum™ 142734
Catalog No. 08-700-250
Add to cart
Description
Description
Spectra/Por Standard Closure Type:
  • Made from polypropylene to aid in sample buoyancy
  • Maintains sample vertical orientation when used with a Spectra/Por Weighted Closure.
  • 5 sealing width sizes: 12 mm, 23mm, 35 mm, 55 mm and 75 mm
  • 4 colors available for easier sample identification
  • Autoclavable
  • 10/pkg; Use with RC dialysis tubing only. Not for use with CE tubing.
Specifications
Specifications
Non-sterile
Autoclavable
10 Pack