Learn More
Diamedix™ 25-Hydroxy Vitamin D Assay
Designed for the quantification of total 25-hydroxy Vitamin D in human serum and plasma
$181.00 - $924.00
Specifications
Certifications/Compliance CE Marked
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Format Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Format Product Type Price Quantity & Availability  
23-451-103
View Documents
Diamedix™
DZ688ACON
2 x 1mL Liquid 25-Hydroxy vitamin D two level control set Pack of 2 for $181.00
Add to cart
23-451-101
View Documents
Diamedix™
DZ688AAPR
96 Tests Lyophilized Automatic plate reader kit Pack of 96 for $924.00
Add to cart
Description
Description
Uses a Vitamin D binding protein which binds Vitamin D3 and D2 at the same affinity.
  • Uses enzyme immunoassay technology (EIA) to measures true total Vitamin D (25-hydroxy vitamin D3 + 25-hydroxy Vitamin D2)
  • Linear from 5.9 to 120.2ng/mL, and has a good correlation with antibody based assays with r2>0.93 and slope of 0.97
  • Has intra and total precisions of <10%
  • Can be run on both manual and automated microplate readers
  • Does not require tedious washing steps between reagents
  • Entire assay procedure takes approximately 2.5 hours
Specifications
Specifications
CE Marked