Diamedix™ 25-Hydroxy Vitamin D Assay

Designed for the quantification of total 25-hydroxy Vitamin D in human serum and plasma

Manufacturer: DIAMEDIX CORPORATION DZ688AAPR

 View more versions of this product

Catalog No. 23-451-101

  Check Availability
Add to Cart

    Specifications

    Product Type Automatic plate reader kit
    For Use With (Equipment) 25-Hydroxy vitamin test kit
    Format Lyophilized
    Includes 1 x 8mL R1, 1 x 7.5mL R2, 1 x 12mL R3, Extraction: 1 x 30mL, Stop: 1 x 8mL, Calibration: 6 x 1mL vial, liquid, Control: 2 x 1mL vial, liquid
    Quantity 96 Tests
    Certifications/Compliance CE Marked

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit