Diamedix™ Aeskulisa™ tTg-A Celiac Disease ELISA
For semiquantitative and qualitative detection of Ig A antibodies against tissue transglutaminase (tTG) in human serum
Manufacturer: DIAMEDIX CORPORATION 307503USREPL
 View more versions of this product
Catalog No. 23-451-126
  Check Availability
Add to Cart
    Specifications
    Product Type tTg-A Celiac Disease ELISA
    Detectable Analytes IgA Antibodies against tissue transglutaminase in Human Serum
    Quantity 4 Pack
  • Description & Specifications
Related Products
Product Content Feedback