Learn More
Diamedix™ Aeskulisa™ tTg-A Celiac Disease ELISA
For semiquantitative and qualitative detection of Ig A antibodies against tissue transglutaminase (tTG) in human serum
$49.75
Specifications
Detectable Analytes IgA Antibodies against tissue transglutaminase in Human Serum
Product Type tTg-A Celiac Disease ELISA
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
23-451-126
View Documents
Diamedix™
307503USREPL
4 Pack
Pack of 4 for $49.75
Add to cart
Description
Description
The assay is an aid in the diagnosis of celiac disease (gluten-sensitive enteropathy) and should be used in conjuction with other serological tests and clinical findings
  • Solid phase enzyme immunoassay
  • Tissue Transglutaminase IgA ELISA used as an aid to diagnose celiac disease
Specifications
Specifications
IgA Antibodies against tissue transglutaminase in Human Serum
tTg-A Celiac Disease ELISA