Learn More
Diamedix™ Aeskulisa™ tTg-G Celiac Disease ELISA
For semiquantitative and qualitative detection of IgG antibodies against tissue transglutaminase (tTG) in human serum
Manufacturer:  DIAMEDIX CORPORATION 307504US
 View more versions of this product
Catalog No. 23-451-125
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
IgG Antibodies against tissue transglutaminase in Human Serum
96 Tests
tTg-G Celiac Disease ELISA