Diamedix™ Dri-Dye Check Strips
For performance verification of microstrip and microplate readers. Diamedix Dri-Dye Check Strips provide easy and economical way to monitor calibration, linearity and repeatability of microstrip and plate readers.
Manufacturer: Diamedix™ 790915
 View more versions of this product
Catalog No. 23-451-052
  Check Availability
Add to Cart
Includes: 6 pre-filled strips per kit, re-usable strip tray (included for use with plate readers), linearity plots, calibration and repeatability charts, instructions
    Specifications
    Product Type Check Strips
    For Use With (Application) For performance verification of microstrip and microplate readers
    Wavelength 405/450nm
    Analytical Method ELISA
    Certifications/Compliance NIST Traceable
    Dri-Dye™ Check Strips
    • Each kit comes with Calibration charts, linearity plots and repeatability records, six prefilled strips and a reusable strip tray for use with plate readers
    • Strips simulate true test conditions better than neutral density filters by providing information about filtering capacity
    • Simply add water, read and compare results to pre-determined acceptance criteria
    • Long shelf life at room temperature
    • NIST traceable absorbance calibration
    • Wavelengths available 405, 450 and 492nm
  • Description & Specifications
Related Products
Product Content Feedback