Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diamedix™ Dri-Dye Check Strips
For performance verification of microstrip and microplate readers. Diamedix Dri-Dye Check Strips provide easy and economical way to monitor calibration, linearity and repeatability of microstrip and plate readers.
$328.00
Specifications
Type | Check Strips |
---|---|
For Use With (Application) | For performance verification of microstrip and microplate readers |
Analytical Method | ELISA |
Certifications/Compliance | NIST Traceable |
Catalog Number | Mfr. No. | Wavelength | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Wavelength | Price | Quantity & Availability | |||||
23-451-050
|
Diamedix™
790900 |
405 nm |
Pack of 6 for $328.00
|
|
|||||
23-451-051
|
Diamedix™
790905 |
450 nm |
Pack of 6 for $328.00
|
|
|||||
Description
Dri-Dye™ Check Strips
- Each kit comes with Calibration charts, linearity plots and repeatability records, six prefilled strips and a reusable strip tray for use with plate readers
- Strips simulate true test conditions better than neutral density filters by providing information about filtering capacity
- Simply add water, read and compare results to pre-determined acceptance criteria
- Long shelf life at room temperature
- NIST traceable absorbance calibration
- Wavelengths available 405, 450 and 492nm
Specifications
Check Strips | |
For performance verification of microstrip and microplate readers | |
ELISA | |
NIST Traceable |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title