Diamedix™ DSX™ and DS2™ Automated ELISA System Software
LIS-Link software for DSX and DS2 to connect to interface.
Manufacturer: Diamedix™ 65088
 View more versions of this product
Catalog No. 23-451-084
  Check Availability
Add to Cart
    Specifications
    For Use With DSX automated ELISA systems
    Product Type LIS-LINK Communication Software
    Analytical Method ELISA
    • Download pending lab orders and upload test results
    • Requires MS-Windows NT 6a or 2000, 5000MHz processor, 64MB memory, 100MB hard drive, video card, CD-ROM, serial port
  • Description & Specifications
Related Products
Product Content Feedback