Diamedix™ Dynex DSX™ Automated ELISA System

The DSX is a fully automated 4-plate processing system capable of performing multiple assays per plate

Manufacturer: DIAMEDIX CORPORATION 65100

 View more versions of this product

Catalog No. 23-451-096

  Check Availability
Add to Cart

    Specifications

    Height (English) 32 in.
    Height (Metric) 80cm
    Hertz 50/60Hz
    Analytical Method ELISA
    Voltage 100 to 240V
    Weight (Metric) 136kg
    Width (English) 42 in.
    Width (Metric) 106cm
    Weight (English) 300 lb.
    Depth (English) 36 in.
    Depth (Metric) 91cm
    Electrical Requirements 100 to 240V 50/60Hz
    Includes No incubator. On-board barcode reader, UPS, Hewlett-Packard PC, keyboard, flat screen monitor and laser print, Diamedix custom rack
    Quantity 96 Samples/Load

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit