Learn More
Diamedix™ Immunetics™ C6 B. burgdorferi (Lyme) ELISA Kit
For use in the presumptive detection of IgG and IgM antibodies to B. burgdorferi in human serum
Manufacturer:  Diamedix™ DKE352096AA
Catalog No. 23-451-129
Add to cart
Description
Description
The assay should be used only on samples from patients with clinical history, signs or symptoms consistent with B. burgdorferi infection, including individuals who have received the licensed recombinant OspA Lyme disease vaccine (Lymerix™)
  • Accurate: Western Blot accuracy in an ELISA
  • Sensitive: More sensitive detection of antibodies in early and late Lyme disease than competing kits
  • Specific: Eliminates vast majority of other ELISA's false positives at the screening step
  • Broadly applicable: Detects infection with all strains of Borrelia which cause Lyme disease, including European strains B. garinii and B. afzelii
  • Convenient: One hour turnaround time, breakaway strips for any number of samples, ready-to-use reagents and controls included
  • Cost-effective: Greatly reduces the number of samples sent for expensive and lengthy confirmatory testing
Specifications
Specifications
Well Plate
Breakaway strips for any number of samples, ready-to-use reagents and control
96 Tests
Infection with all strains of Borrelia which cause Lyme disease, including European strains B. garinii and B. afzelii
Immunetics™ C6 B. burgdorferi (Lyme) ELISA kit