Learn More
Diamedix™ Immunosimplicity™ H. pylori IgG Test Kit
SDP
Detects IgG antibodies to Helicobacter pylori antigen in human sera
Manufacturer:  Diamedix™ 720130
Catalog No. 23-451-035
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
  • Handle samples, calibrator, controls and the materials that contact them as potential biohazards
  • Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide
Includes: Antigen Wells, Cut-off Calibrator, Positive Control, Negative Control, Sample A Diluent, Wash T Concentrate (20X), Conjugate, Substrate HRP, Stop N Solution
Description
Description
  • Qualitative indirect enzyme immunoassay
  • Used as an aid in diagnosis of H. pylori infection in adult patients with gastrointestinal symptoms
  • For in vitro diagnostic use
Reagent volumes/configurations are suitable for manual use, automated Mago™ systems and Dynex DSX™ and DS2™ systems
Specifications
Specifications
96 Tests
H. pylori Antigen in Human sera
Antigen wells, cut-off calibrator, positive control, negative control, sample A diluent, 20X wash T concentrate, conjugate, substrate HRP, stop N solution
H. pylori IgG Test Kit