Diamedix™ Immunosimplicity™ Rubella IgM Capture Test Kit
Detects IgM antibodies to rubella in human serum
Manufacturer: DIAMEDIX CORPORATION 300371A
 View more versions of this product
Catalog No. 23-451-041
  Check Availability
Add to Cart
Alert
Handle samples, standards, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
    Specifications
    Product Type Immunosimplicity™ Rubella IgM Capture Antigen
    Detectable Analytes IgM Antibodies to Rubella
    Includes Anti-IgM coated wells, cut-off calibrator, low positive control, negative control, lyophilized agent, 30X Tracer, tracer diluent, sample A diluent, wash S concentrate 20X, substrate G, stop M solution
    Packaging Vial
    Quantity 3 Vials
  • Description & Specifications
Related Products
Product Content Feedback