Learn More
Diamedix™ Immunosimplicity™ Rubella IgM Capture Test Kit
Detects IgM antibodies to rubella in human serum
Manufacturer:  DIAMEDIX CORPORATION 720370
 View more versions of this product
Catalog No. 23-451-024
  Check Availability
Add to cart
Alert:
Handle samples, standards, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.
Specifications Description & Specifications
Specifications
IgM Antibodies to Rubella
Anti-IgM coated wells, cut-off calibrator, low positive control, negative control, lyophilized agent, 30X Tracer, tracer diluent, sample A diluent, wash S concentrate 20X, substrate G, stop M solution
96 Tests
Immunosimplicity™ Rubella IgG Test Kit