Diamedix™ Immunosimplicity™ Toxoplasma IgM Capture Test Kit

Detects IgM antibodies to Toxoplasma gondii in human serum

Manufacturer: DIAMEDIX CORPORATION 300312A

 View more versions of this product

Catalog No. 23-451-040

  Check Availability
Add to Cart

Alert

Handle samples, calibrator, controls and the materials that contact them as potential biohazards.
Certain reagents contain the preservatives ProClin™ 300 or Sodium Azide.


    Specifications

    Product Type Immunosimplicity™ Toxoplasma IgM Capture Antigen
    Detectable Analytes IgM Antibodies
    For Use With (Application) Detects IgM antibodies to Toxoplasma gondii in human serum
    Format Lyophilized
    Quantity 3 Vials

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit