Diamedix™ MAGO™ 4S Automated Immunoassay System
Simultaneous ELISA and IFA test processing doubles result output and increases efficiency
Manufacturer: DIAMEDIX CORPORATION 251200
 View more versions of this product
Catalog No. 23-451-093
  Check Availability
Add to Cart
    Specifications
    Product Type Automated Immunoassay Analyzer
    Analytical Method ELISA/IFA
    Weight (Metric) 90.9kg
    Weight (English) 200 lb.
    For Use With (Application) ELISA and IFA testing
    Quantity 120 Samples/Load
  • Description & Specifications
Related Products
Product Content Feedback