Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamedix™ Phoenix Bio-Tech™ TREP-SURE™ Anti-Treponema EIA Screen

For in vitro diagnostic detection of Treponema pallidum (syphilis) antibodies in human serum or EDTA and citrated plasma

Manufacturer:  Diamedix™ TS960

 View more versions of this product

Catalog No. 23-451-131


Add to cart
This item is not returnable. View return policy

Description

Description

  • Qualitative enzyme immunoassay
  • Total test incubation time: 105 minutes
  • Package size: 96 tests, 960 tests, 1920 tests
  • Required sample size: 100µL

Unopened kits are stable through the expiration date stated on the label expiry date (or 30 days once opened).

Specifications

Specifications

Treponema pallidum Antibodies in Human Serum or EDTA and citrated Plasma
Microwell 12 x 8-well strips, positive Control 1.8mL/vial, cut-off calibrator 1.8ml/ vial, negative control 1.8mL/vial, conjugate 13mL, Wash Buffer 50mL, substrate 13mL, stop solution 13mL, ten 96 well plates
Trep-Sure™ Anti-Treponema EIA Screen
96 Tests
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit