Learn More
Diamedix™ Phoenix Bio-Tech™ TREP-SURE™ Anti-Treponema EIA Screen
For in vitro diagnostic detection of Treponema pallidum (syphilis) antibodies in human serum or EDTA and citrated plasma
Manufacturer:  Diamedix™ TS96E
 View more versions of this product
Catalog No. 23-451-132
Add to cart
Description
Description
  • Qualitative enzyme immunoassay
  • Total test incubation time: 105 minutes
  • Package size: 96 tests, 960 tests, 1920 tests
  • Required sample size: 100µL
Unopened kits are stable through the expiration date stated on the label expiry date (or 30 days once opened).
Specifications
Specifications
Treponema pallidum Antibodies in Human Serum or EDTA and citrated Plasma
Extra reagents for TS 96
Trep-Sure™ ELISA Extra Reagent
96 Tests