Learn More
Diamedix™ Phoenix Bio-Tech™ TREP-SURE™ Anti-Treponema EIA Screen
For in vitro diagnostic detection of Treponema pallidum (syphilis) antibodies in human serum or EDTA and citrated plasma
Manufacturer:  Diamedix™ TS960E
 View more versions of this product
Catalog No. 23-451-133
Add to cart
This item is not returnable. View return policy
Description
Description
  • Qualitative enzyme immunoassay
  • Total test incubation time: 105 minutes
  • Package size: 96 tests, 960 tests, 1920 tests
  • Required sample size: 100µL
Unopened kits are stable through the expiration date stated on the label expiry date (or 30 days once opened).
Specifications
Specifications
Treponema pallidum Antibodies in Human Serum or EDTA and citrated Plasma
Extra reagents for TS 960
Trep-Sure™ ELISA Extra Reagent
960 Tests