Learn More
Diamedix™ Phoenix Bio-Tech™ TREP-SURE™ Anti-Treponema EIA Screen
For in vitro diagnostic detection of Treponema pallidum (syphilis) antibodies in human serum or EDTA and citrated plasma
$461.00 - $3720.00
Specifications
Detectable Analytes Treponema pallidum Antibodies in Human Serum or EDTA and citrated Plasma
View More Specs
Products 4
Catalog Number Mfr. No. Quantity Includes Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Includes Product Type Price Quantity & Availability  
23-451-132
View Documents
Diamedix™
TS96E
96 Tests Extra reagents for TS 96 Trep-Sure™ ELISA Extra Reagent
Pack of 96 for $480.00
Add to cart
23-451-133
View Documents
Diamedix™
TS960E
960 Tests Extra reagents for TS 960 Trep-Sure™ ELISA Extra Reagent
Pack of 960 for $3,720.00
Add to cart
23-451-130
View Documents
Diamedix™
TS96
960 Tests Microwell 120 x 8-well strips, positive control 1.8mL x 3 vials, cut-off calibrator 1.8mL x 4 vials, negative control 1.8mL x 3 vials, conjugate 5 x 25mL, Wash Buffer 500mL, substrate 5 x 25mL, stop solution 5 x 25mL, one 96 well plate Trep-Sure™ Anti-Treponema EIA Screen
Pack of 96 for $461.00
Add to cart
23-451-131
View Documents
Diamedix™
TS960
96 Tests Microwell 12 x 8-well strips, positive Control 1.8mL/vial, cut-off calibrator 1.8ml/ vial, negative control 1.8mL/vial, conjugate 13mL, Wash Buffer 50mL, substrate 13mL, stop solution 13mL, ten 96 well plates Trep-Sure™ Anti-Treponema EIA Screen
Pack of 960 for $3,500.00
Add to cart
Description
Description
  • Qualitative enzyme immunoassay
  • Total test incubation time: 105 minutes
  • Package size: 96 tests, 960 tests, 1920 tests
  • Required sample size: 100µL
Unopened kits are stable through the expiration date stated on the label expiry date (or 30 days once opened).
Specifications
Specifications
Treponema pallidum Antibodies in Human Serum or EDTA and citrated Plasma