Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diaminomaleonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
1187-42-4
C4H4N4
108.1
MFCD00001870
DPZSNGJNFHWQDC-ARJAWSKDSA-N
diaminomaleonitrile,2,3-diaminomaleonitrile,hydrogen cyanide tetramer,maleonitrile, diamino,ccris 921,2-butenedinitrile, 2,3-diamino-, 2z,2z-diaminobut-2-enedinitrile,2,3-diamino maleonitrile,2-butenedinitrile, 2,3-diamino-, z,z-2,3-diaminobut-2-enedinitrile
2723951
(Z)-2,3-diaminobut-2-enedinitrile
C(#N)C(=C(C#N)N)N
1187-42-4
C4H4N4
108.1
MFCD00001870
DPZSNGJNFHWQDC-ARJAWSKDSA-N
diaminomaleonitrile,2,3-diaminomaleonitrile,hydrogen cyanide tetramer,maleonitrile, diamino,ccris 921,2-butenedinitrile, 2,3-diamino-, 2z,2z-diaminobut-2-enedinitrile,2,3-diamino maleonitrile,2-butenedinitrile, 2,3-diamino-, z,z-2,3-diaminobut-2-enedinitrile
2723951
(Z)-2,3-diaminobut-2-enedinitrile
C(#N)C(=C(C#N)N)N
Color Brown
Melting Point 178.0°C to 179.0°C
Quantity 500g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula NCC(NH2)=C(NH2)CN
Beilstein 04, II, 949
Fieser 05,175; 04,480; 13,327; 01,1256; 02,454; 04,565; 06,657; 07,414; 08,524; 09,512; 10,455; 13,301
Solubility Information Solubility in water: 5g/L (25°C). Other solubilities: soluble in methanol, ethanol, dmf and dimethyl, sulfoxide, slightly soluble in ethyl ether and dioxane, insol. in benzene,acetone,xylene,toluene and cs2
Formula Weight 108.1
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Diaminomaleonitrile
Show More Show Less

GHS H Statement
Toxic if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Contact with acids liberates very toxic gas.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

EINECSNumber : 214-697-6