Alfa Aesar™ Diamminepalladium(II) nitrite solution, Pd 8-10% w/w (cont. Pd)

Manufacturer: Alfa Aesar™ 11038NA

Quantity 2g

 View more versions of this product

Catalog No. AA11038NA

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 28068-05-5
    Formula Weight 232.47 (Anhydrous)
    MDL Number MFCD00038712
    UN Number UN2672
    Synonym Diamminedinitritopalladate(II)
    Chemical Name or Material Diamminepalladium(II) nitrite solution

    Specifications

    Formula Weight 232.47 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P310-P301+P312-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Health Hazard 2 GHS H Statement
    H318-H302
    Causes serious eye damage.
    Harmful if swallowed.
    MDL Number MFCD00038712
    CAS 28068-05-5
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMMONIA SOLUTION
    Odor Ammonia-like
    Synonym Diamminedinitritopalladate(II)
    Concentration Pd 8 to 10% w/w (cont. Pd)
    Quantity 2g
    TSCA TSCA
    UN Number UN2672
    Color Yellow
    Chemical Name or Material Diamminepalladium(II) nitrite solution

    GHS H Statement
    H318-H302
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P280-P264-P305+P351+P338-P310-P301+P312-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit