Learn More
Globe Scientific Diamond RV-Pette PRO™ Dispenser Syringe Tips
For use with Diamond Rv-Pette PRO™ Repeat Volume Pipettor.
Supplier: Globe Scientific 3926
This item is not returnable.
View return policy
Description
- Standard color coding ensures easy volume identification
- Safe, secure choice for handling a wide range of liquids including those that are viscous, infectious, and volatile
- Tested during the automated manufacturing process
- Available in 12 sizes for dispensing volumes from 1μL to 5500μL
- Available non-sterile and sterile
- Sterile tips are individually wrapped and certified to be RNase, DNase, ATP, and pyrogen free for most critical applications
Specifications
2.5 mL | |
Non-sterile | |
Single-use | |
No | |
Polypropylene/Polyethylene |
Non-Filtered | |
Natural | |
Universal Fit Repeater Pipettor | |
Diamond RV-Pette PRO |