Learn More
DIAPH2 Monoclonal Antibody (V78 P3C10-D3), APC, Invitrogen™
Mouse Monoclonal Antibody
Supplier:  Thermo Scientific MA545918
Encompass
Catalog No. PIMA545918
Add to Cart
This item is not returnable. View return policy
Description
Description
DIAPH2 Monoclonal antibody specifically detects DIAPH2 in Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DIAPH2
Monoclonal
1 mg/mL
2.15mM MES, 47.35mM phosphate with 1.033mM EDTA and no preservative; pH 7.4
O70566
DIAPH2
Synthetic peptide of Human DIAPH2
100 μg
Primary
Mouse, Rat
Liquid
ELISA, Immunohistochemistry, Western Blot
V78 P3C10-D3
APC
DIAPH2
DIA; DIA2; Dia3; Diap2; Diaph2; diaphanous homolog 2; diaphanous homolog 2 (Drosophila); diaphanous related formin 2; diaphanous-related formin 2; diaphanous-related formin-2; diaphorase-2; DRF2; E430022I22Rik; FLJ11167; mDia3; mDia3 isoform; POF; POF2; Protein diaphanous homolog 2; RGD1564399; RP11-212B22.1
Mouse
Protein G
RUO
317189, 54004
4° C
IgG1 κ
Product Suggestions
Product Suggestions