Learn More
Abnova™ Diaphorase (Cl. Kluyveri), NAD & NADP Removed
Used for Enzyme
Manufacturer:  Abnova™ P5286
Catalog No. 89-941-977
Add to cart
Description
Description
#REF!
Specifications
Specifications
Q97E86
Lyophilized
Native protein purified from Cl. kluyveri
Store at 4°C. For long term storage store at -20°C. Avoid exposure to light after reconstitution.
DIAPHORASE
Native protein purified from Cl. kluyveri
Clostridium Species
Enzyme Activity
Diaphorase (Cl. Kluyveri), NAD & NADP Removed
2000 Units
RUO
Diaphorase Assay
≥25 units per mg dry weight. One Unit reduces 1 umole of DCPIP per minute at 25°C, pH 8.5.
None