Learn More
DiaSorin Arrow Extraction Kits
Supports fully automated extraction and cell separation using Arrow instrument
Manufacturer:  DiaSorin Inc 12.01.02
 View more versions of this product
Catalog No. 23-111-403
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Creates pure Nucleic Acids in the tube
DiaSorin arrow automated cell separator
Arrow BUGS'n BEADS™ (CE, IVD)
96 Preps