Learn More
DiaSorin Bullet PRO Automated Nucleic Acid Extraction Instrument
Fully automated and effective sample preparation instrument for molecular diagnostics
Manufacturer:  DiaSorin 8.11.04
Catalog No. 23-111-406
Add to cart
Description
Description
Magnetic bead-based chemistry for the extraction of nucleic acids from the most difficult samples and for the most demanding molecular applications.
  • Extraction from primary sample tubes to PCR set-up, is done in about 2 hours, depending on the protocol
  • Extracts up to 96 samples with minimal hands-on time
  • Can be adapted to a variety of downstream amplification methods
Sample types for nucleic acid extraction:
  • Swabs
  • Blood
  • Sputum
  • Urine
  • Serum
  • Plasma
  • Culture
  • Stool
Only use the transformer supplied with the instrument or one that has been recommended by DiaSorin or your DiaSorin distributor.
For high throughput applications
Specifications
Specifications
High throughput applications
Swabs, blood, sputum, urine, serum, plasma, culture, stool
17.5 in.
Touch Screen
51cm
17.4 in.
48.5 lb.
22kg
47cm
18.3 in.
96.0