Learn More
StatLab DIASTASE PAS GLY DIGESTN 50 GM
SDP
Supplier:  StatLab AHDIA50
Encompass_Preferred
Used to detemine if glycogen is present in tissue sections.
Catalog No. NC9033522
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.