Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diatomaceous Earth, J.T.Baker™

Click to view available options
Quantity:
500g
1kg
2.5kg
3kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle
68855-54-9
CNa2O3
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
68855-54-9
CNa2O3
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
Quantity 3kg
Specific Gravity 1L = 2.30kg
Particle Size 33 Micron
Packaging Poly Pail
Chemical Name or Material Diatomaceous Earth