Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diatrizoate Sodium, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
737-31-5
C11H8I3N2NaO4
635.90
ZEYOIOAKZLALAP-UHFFFAOYSA-M
sodium 3,5-diacetamido-2,4,6-triiodobenzoate
[Na+].CC(=O)NC1=C(I)C(C([O-])=O)=C(I)C(NC(C)=O)=C1I
737-31-5
C11H8I3N2NaO4
635.90
ZEYOIOAKZLALAP-UHFFFAOYSA-M
sodium 3,5-diacetamido-2,4,6-triiodobenzoate
[Na+].CC(=O)NC1=C(I)C(C([O-])=O)=C(I)C(NC(C)=O)=C1I

S1263, 737-31-5, C11H8I3N2NaO4

Spectrum™ Diatrizoate Sodium, USP is used as a radiocontrast agent for imaging the gastrointestinal tract in patients allergic to barium. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Percent Purity 98 to 102%
Packaging Amber Glass Bottle
Quantity 25 g