Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Diazoxide, 98%

Catalog Number AAJ6601003
Click to view available options
Quantity:
100 mg
500 mg
1 g
364-98-7
364-98-7

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Description White to off-white
Content And Storage 2°C to 8°C
Molecular Formula C8H7ClN2O2S
Formula Weight 230.67
Synonym 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide;
Melting Point 330°C to 331°C
Solubility Soluble in 0.1M NaOH. Insoluble in water or in methanol.
Form Powder
Assay 98%
EINECSNumber 206-668-1
Recommended Storage Keep cold

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit