Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazoxide, USP, 97-102%, Spectrum™

Click to view available options
Quantity:
1 kg
Packaging:
Amber Glass Bottle

D1145, 364-98-7, C8H7ClN2O2S

Spectrum™ Diazoxide, USP is a vasodilator is used to treat very low blood sugar levels (hypoglycemia). All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.001
Quantity 1 kg
Loss on Drying 0.005
CAS 364-98-7
CAS Max % 1
Molecular Formula C8H7ClN2O2S
InChI Key GDLBFKVLRPITMI-UHFFFAOYSA-N
SMILES CC1=NS(=O)(=O)C2=CC(Cl)=CC=C2N1
IUPAC Name 7-chloro-3-methyl-4H-1λ⁶,2,4-benzothiadiazine-1,1-dione
Molecular Weight (g/mol) 230.67
Percent Purity 97 to 102%
Show More Show Less