Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibasic Calcium Phosphate, Dihydrate, Powder, USP, 98-105%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7789-77-7
CaH5O6P
172.09
XAAHAAMILDNBPS-UHFFFAOYSA-L
calcium dihydrate hydrogen phosphate
O.O.[Ca++].OP([O-])([O-])=O
7789-77-7
CaH5O6P
172.09
XAAHAAMILDNBPS-UHFFFAOYSA-L
calcium dihydrate hydrogen phosphate
O.O.[Ca++].OP([O-])([O-])=O

CA160, 7789-77-7, CaHPO4•2H2O

Spectrum™ Dibasic Calcium Phosphate, Dihydrate, Powder, USP is used as a dietary supplement. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 3 μg/g
Chloride 0.0025
Percent Purity 98 to 105%
Packaging Poly Pail
Quantity 12 kg